This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IL6ST SIRNA Assays ›
Gene Symbol:
Il6st
Gene Name:
interleukin 6 signal transducer
Gene Aliases:
Ac1055, Gp130, Il-6rb
Chromosome Location:
Chr.2: 44279206 - 44319427 on Build Rnor_6.0
UniGene ID:
Rn.12138
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218858

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Il6st
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008725.3 NP_001008725.2 5 809
XM_006231928.2 XP_006231990.1 7 1062
XM_008760736.1 XP_008758958.1 7 1907
GenBank mRNA AY310138.1 - 779
JQ013730.1 - 5 1160
M92340.1 - 5 843

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_175767, NM_002184, NM_001190981, AB015706, M57230, and others.
This siRNA also targets the following transcripts in Mouse: NM_010560, X62646, M83336, BC020454, BC058679.

Gene Ontology Categories:

Function(s) Process(es)

glycogen metabolic process
triglyceride mobilization
signal transduction
response to nutrient
positive regulation of cell proliferation
regulation of Notch signaling pathway
positive regulation of smooth muscle cell migration
cytokine-mediated signaling pathway
response to cytokine
interleukin-11-mediated signaling pathway
oncostatin-M-mediated signaling pathway
positive regulation of T cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
positive regulation of tyrosine phosphorylation of Stat3 protein
positive regulation of osteoblast differentiation
regulation of hormone secretion
positive regulation of astrocyte differentiation
leukemia inhibitory factor signaling pathway
negative regulation of cytosolic calcium ion concentration
interleukin-6-mediated signaling pathway
negative regulation of interleukin-6-mediated signaling pathway
interleukin-27-mediated signaling pathway
ciliary neurotrophic factor-mediated signaling pathway
ciliary neurotrophic factor receptor activity
interleukin-6 receptor activity
interleukin-11 receptor activity
leukemia inhibitory factor receptor activity
oncostatin-M receptor activity
ciliary neurotrophic factor receptor binding
interleukin-6 receptor binding
growth factor binding
interleukin-11 binding
interleukin-6 binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0