This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MGC105649 SIRNA Assays ›
Gene Symbol:
MGC105649
Gene Name:
hypothetical LOC302884
Gene Aliases:
Nmes1
Chromosome Location:
Chr.3: 114772603 - 114776158 on Build Rnor_6.0
UniGene ID:
Rn.106101
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218766

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MGC105649
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008518.2 NP_001008518.1 2, 3 94
XM_008762164.1 XP_008760386.1 1, 2 185
GenBank mRNA BC086337.1 - 1, 2 70 7301704
CB585387.1 - 2, 3 99
FQ216036.1 - 1, 2 191
FQ221053.1 - 2, 3 91
FQ223264.1 - 2, 3 90

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001004174, AK141297, BC049633, AK150561.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0