This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM64 SIRNA Assays ›
Gene Symbol:
TMEM64
Gene Name:
transmembrane protein 64
Gene Aliases:
-
Chromosome Location:
Chr.8: 90621995 - 90645905 on Build GRCh38
UniGene ID:
Hs.567759
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 218719

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM64
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008495.3 NP_001008495.2 1 768
NM_001146273.1 NP_001139745.1 1 768
GenBank mRNA AK123958.1 - 1 462
AK300583.1 - 1 375
AL834364.1 - 1 474
AY147881.1 - 1 573

Back To Top

More Information


Gene Ontology Categories:

Function(s)

regulation of ATPase activity
canonical Wnt signaling pathway involved in osteoblast differentiation
positive regulation of fat cell differentiation
negative regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
positive regulation of bone resorption
regulation of cytosolic calcium ion concentration
negative regulation of canonical Wnt signaling pathway

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0