This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNOT8 SIRNA Assays ›
Gene Symbol:
Cnot8
Gene Name:
CCR4-NOT transcription complex, subunit 8
Gene Aliases:
-
Chromosome Location:
Chr.10: 43538155 - 43551271 on Build Rnor_6.0
UniGene ID:
Rn.203142
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218642

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnot8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008382.1 NP_001008383.1 6 770
XM_006246392.2 XP_006246454.1 5 829
GenBank mRNA BC085856.1 - 6 770 7190112

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, deadenylation-dependent decay
transcription, DNA-templated
biological_process
positive regulation of cell proliferation
exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
positive regulation of mRNA catabolic process
RNA phosphodiester bond hydrolysis, exonucleolytic
3'-5'-exoribonuclease activity
molecular_function
nucleic acid binding
poly(A)-specific ribonuclease activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0