This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POMGNT1 SIRNA Assays ›
Gene Symbol:
Pomgnt1
Gene Name:
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
Gene Aliases:
-
Chromosome Location:
Chr.5: 135007343 - 135017220 on Build Rnor_6.0
UniGene ID:
Rn.162088
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 218254

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pomgnt1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007747.1 NP_001007748.1 19 1778
XM_006238683.2 XP_006238745.1 18 1885
GenBank mRNA BC083641.1 - 19 1778 7134894

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_026651, NM_029786, AK178221, AK180366, AK210391, and others.

Gene Ontology Categories:

Function(s) Process(es)

protein O-linked glycosylation
biological_process
molecular_function
beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0