This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PIGQ SIRNA Assays ›
Gene Symbol:
Pigq
Gene Name:
phosphatidylinositol glycan anchor biosynthesis, class Q
Gene Aliases:
-
Chromosome Location:
Chr.10: 15289530 - 15305595 on Build Rnor_6.0
UniGene ID:
Rn.12777
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 217870

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pigq
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007607.1 NP_001007608.1 11 1902
XM_006245942.2 XP_006246004.1 11 1981
XM_008767559.1 XP_008765781.1 11 1921
GenBank mRNA BC081875.1 - 11 1902 7107681

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: AF030178.

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

GPI anchor biosynthetic process
phosphatidylinositol N-acetylglucosaminyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0