This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GSTO1 SIRNA Assays ›
Gene Symbol:
Gsto1
Gene Name:
glutathione S-transferase omega 1
Gene Aliases:
SPG-R
Chromosome Location:
Chr.1: 267607437 - 267617387 on Build Rnor_6.0
UniGene ID:
Rn.25166
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 217859

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gsto1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007602.1 NP_001007603.1 2 173
GenBank mRNA AB008807.1 - 2 155
BC079363.1 - 2 173 7109248
FJ179405.1 - 2 91
FQ218105.1 - 2 148
FQ220742.1 - 2 115

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
L-ascorbic acid metabolic process
L-ascorbic acid biosynthetic process
xenobiotic catabolic process
oxidation-reduction process
negative regulation of ryanodine-sensitive calcium-release channel activity
positive regulation of ryanodine-sensitive calcium-release channel activity
cellular response to arsenic-containing substance
glutathione transferase activity
oxidoreductase activity
glutathione dehydrogenase (ascorbate) activity
methylarsonate reductase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0