This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP251 SIRNA Assays ›
Gene Symbol:
Zfp251
Gene Name:
zinc finger protein 251
Gene Aliases:
9130001M19Rik, AV339307
Chromosome Location:
Chr.15: 76852131 - 76871435 on Build GRCm38
UniGene ID:
Mm.38195
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 217789

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp251
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007568.2 NP_001007569.1 4 3220
GenBank mRNA AK018584.1 - 381
AK047299.1 - 496
AK140082.1 - 4 3211
AK153967.1 - 3 3060
BC041112.1 - 1657
BC059071.1 - 4 3220 6826152

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0