This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ADAM22 SIRNA Assays ›
Gene Symbol:
Adam22
Gene Name:
a disintegrin and metallopeptidase domain 22
Gene Aliases:
2900022I03Rik, AI854032, MDC2
Chromosome Location:
Chr.5: 8072348 - 8368268 on Build GRCm38
UniGene ID:
Mm.275895
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 217697

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adam22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007220.2 NP_001007221.1 21 1829
NM_001007221.2 NP_001007222.1 21 1829
NM_001098225.1 NP_001091695.1 21 1829
GenBank mRNA AB009674.1 - 21 1825
BC117707.1 - 21 1699
BC117708.1 - 21 1701
HM004095.1 - 21 1702

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008762748.

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
adult locomotory behavior
Schwann cell differentiation
myelination in peripheral nervous system
gliogenesis
metalloendopeptidase activity
protein binding
metallopeptidase activity
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0