siRNA Details
Gene Symbol: Drc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007009.1 | NP_001007010.1 | 7, 8 | 929 | ||
GenBank mRNA | BC083825.1 | - | 7, 8 | 929 | 7112171 |

Gene Symbol: Drc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007009.1 | NP_001007010.1 | 7, 8 | 929 | ||
GenBank mRNA | BC083825.1 | - | 7, 8 | 929 | 7112171 |