This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DRC1 SIRNA Assays ›
Gene Symbol:
Drc1
Gene Name:
dynein regulatory complex subunit 1
Gene Aliases:
Ccdc164
Chromosome Location:
Chr.6: 27425237 - 27460038 on Build Rnor_6.0
UniGene ID:
Rn.51279
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 217644

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Drc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007009.1 NP_001007010.1 2 262
GenBank mRNA BC083825.1 - 2 262 7112171

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_145038, AK289953, AK307094, AK057222, BC009488, and others.

Gene Ontology Categories:

Function(s)

cilium-dependent cell motility
axonemal dynein complex assembly
bacterial-type flagellum-dependent cell motility

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0