This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ECSIT SIRNA Assays ›
Gene Symbol:
Ecsit
Gene Name:
ECSIT signalling integrator
Gene Aliases:
-
Chromosome Location:
Chr.8: 23085598 - 23099062 on Build Rnor_6.0
UniGene ID:
Rn.8455
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 217582

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ecsit
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006986.1 NP_001006987.1 7 1072
XM_006242636.2 XP_006242698.1 7 1137
XM_006242637.2 XP_006242699.1 7 1126
XM_006242638.2 XP_006242700.1 7 1107
GenBank mRNA BC083762.1 - 7 1072 7193076
FQ209658.1 - 7 1080
FQ212584.1 - 7 1103
FQ215401.1 - 7 1101

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mesoderm formation
regulation of transcription from RNA polymerase II promoter
transmembrane receptor protein serine/threonine kinase signaling pathway
BMP signaling pathway
innate immune response
regulation of oxidoreductase activity
oxidation-reduction process
transcription factor activity, sequence-specific DNA binding
signal transducer activity
oxidoreductase activity, acting on NAD(P)H

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0