This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CXCL17 SIRNA Assays ›
Gene Symbol:
CXCL17
Gene Name:
C-X-C motif chemokine ligand 17
Gene Aliases:
DMC, Dcip1, UNQ473, VCC-1, VCC1
Chromosome Location:
Chr.19: 42428538 - 42443048 on Build GRCh38
UniGene ID:
Hs.445586
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 217293

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CXCL17
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198477.2 NP_940879.1 3 467
NR_133910.1 3 484
GenBank mRNA AK292985.1 - 3 365
AY358433.1 - 3 403
AY598464.1 - 3 467
BC093946.1 - 3 318 7939791
BC112095.1 - 3 320 8327563

Back To Top

More Information


Panther Classification:

Molecular Function -

chemokine cytokine intercellular signal molecule

Gene Ontology Categories:

Function(s)

angiogenesis
chemotaxis
cell differentiation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0