This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LRRC8A SIRNA Assays ›
Gene Symbol:
LRRC8A
Gene Name:
leucine rich repeat containing 8 family member A
Gene Aliases:
AGM5, LRRC8, SWELL1
Chromosome Location:
Chr.9: 128882112 - 128918042 on Build GRCh38
UniGene ID:
Hs.643600
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 217267

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LRRC8A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127244.1 NP_001120716.1 3 1743
NM_001127245.1 NP_001120717.1 2 1367
NM_019594.3 NP_062540.2 3 1474
XM_005252095.2 XP_005252152.1 3 1604
XM_005252096.3 XP_005252153.1 3 2767
XM_006717186.2 XP_006717249.1 4 1819
XM_011518863.1 XP_011517165.1 3 1410
XM_011518864.2 XP_011517166.1 4 1528
XM_011518865.2 XP_011517167.1 2 1246
GenBank mRNA AB037858.1 - 2 1283
AK024649.1 - 2 1304
AK027495.1 - 1 410
AK074723.1 - 1 446
AK289842.1 - 3 1743
AY143166.1 - 1 1220
AY358286.1 - 1 507
BC051322.1 - 3 1386 6250713

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001024782, XM_006233814, XM_008761660, BC097371.
This siRNA also targets the following transcripts in Mouse: NM_177725.

Gene Ontology Categories:

Function(s) Process(es)

pre-B cell differentiation
anion transport
cell volume homeostasis
response to osmotic stress
regulation of anion transport
anion transmembrane transport
volume-sensitive anion channel activity
anion channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0