This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CARHSP1 SIRNA Assays ›
Gene Symbol:
CARHSP1
Gene Name:
calcium regulated heat stable protein 1
Gene Aliases:
CRHSP-24, CRHSP24, CSDC1
Chromosome Location:
Chr.16: 8852942 - 8869012 on Build GRCh38
UniGene ID:
Hs.632184
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215843

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CARHSP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042476.2 NP_001035941.1 4 784
NM_001278260.1 NP_001265189.1 5 592
NM_001278261.1 NP_001265190.1 5 731
NM_001278262.1 NP_001265191.1 5 690
NM_001278263.1 NP_001265192.1 4 466
NM_001278264.1 NP_001265193.1 4 608
NM_001278265.1 NP_001265194.1 4 600
NM_001278266.1 NP_001265195.1 4 576
NM_014316.3 NP_055131.2 4 478
XM_005255228.4 XP_005255285.3 4 642
XM_005255229.4 XP_005255286.1 4 454
XM_011522444.2 XP_011520746.1 4 453
GenBank mRNA AF115345.1 - 3 435
AK311777.1 - 4 472
BC001333.1 - 4 784
BC003366.1 - 4 475 2900952
BC108283.1 - 4 458 4430943
BM920489.1 - 4 466
CA396216.1 - 4 535
HY090855.1 - 4 478

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
intracellular signal transduction
regulation of mRNA stability
DNA binding
mRNA 3'-UTR binding
protein binding
phosphatase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0