This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF593 SIRNA Assays ›
Gene Symbol:
ZNF593
Gene Name:
zinc finger protein 593
Gene Aliases:
ZT86
Chromosome Location:
Chr.1: 26169528 - 26170873 on Build GRCh38
UniGene ID:
Hs.477273
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215798

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF593
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015871.4 NP_056955.2 3 484
XM_017001398.1 XP_016856887.1 2 947
GenBank mRNA BC002580.2 - 3 462 3143091
BC019267.1 - 3 457 2823658
D45213.1 - 3 393
DA124660.1 - 3 484
U38444.1 - 1 73

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
mitophagy in response to mitochondrial depolarization
DNA binding
transcription corepressor activity
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0