This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other JTB SIRNA Assays ›
Gene Symbol:
JTB
Gene Name:
jumping translocation breakpoint
Gene Aliases:
HJTB, HSPC222, PAR, hJT
Chromosome Location:
Chr.1: 153974269 - 153977975 on Build GRCh38
UniGene ID:
Hs.6396
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 215687

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: JTB
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006694.3 NP_006685.1 2, 3 852
GenBank mRNA AB016488.1 - 2, 3 561
AF115850.2 - 2, 3 550
AF131797.1 - 2, 3 433
AF151056.1 - 2 608
AK312106.1 - 2, 3 534
BC000499.1 - 2, 3 574 2820861
BC000996.2 - 2, 3 501 2901221
BC001363.1 - 2, 3 539 3050800
BC001667.1 - 2, 3 504 2823348
BC004239.1 - 3 561 3626047
BC019277.1 - 2, 3 495 2823348
BM907938.1 - 613
BT007285.1 - 2, 3 129
CR456985.1 - 2, 3 129

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
mitotic nuclear division
apoptotic mitochondrial changes
regulation of cell proliferation
positive regulation of protein kinase activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0