This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OR2J2 SIRNA Assays ›
Gene Symbol:
OR2J2
Gene Name:
olfactory receptor family 2 subfamily J member 2
Gene Aliases:
OR6-19, OR6-8, OR6.3.8, ORL684, dJ80I19.4, hs6M1-6
Chromosome Location:
Chr.6: 29173534 - 29174574 on Build GRCh38
UniGene ID:
Hs.163518
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 214346

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: OR2J2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030905.2 NP_112167.2 1 793
GenBank mRNA BC069121.1 - 1 691 7216975
BC108896.1 - 1 694 40003949
BC108897.1 - 1 691 40003955
BC108898.1 - 1 691 40003956
BC108899.1 - 1 691 40003957

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
G-protein coupled receptor activity
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0