This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FCAR SIRNA Assays ›
Gene Symbol:
FCAR
Gene Name:
Fc fragment of IgA receptor
Gene Aliases:
CD89, CTB-61M7.2, FcalphaRI
Chromosome Location:
Chr.19: 54874093 - 54891420 on Build GRCh38
UniGene ID:
Hs.659872
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 213341

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FCAR
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002000.3 NP_001991.1 1 220
NM_133269.3 NP_579803.1 1 220
NM_133271.3 NP_579805.1 1 220
NM_133272.3 NP_579806.1 1 220
NM_133273.3 NP_579807.1 1 220
NM_133274.3 NP_579808.1 1 220
NM_133277.3 NP_579811.1 1 220
NM_133278.3 NP_579812.1 1 220
XM_011526625.2 XP_011524927.1 1 81
GenBank mRNA AK290922.1 - 1 78
BC027953.1 - 1 47 5211453
BX643574.1 - 1 65
CV029531.1 - 1 23
D87853.1 - 1 23
D87854.1 - 1 23
D87855.1 - 1 23
D87856.1 - 1 23
D87857.1 - 1 23
D87858.1 - 1 23
D87859.1 - 1 23
D87860.1 - 1 23
D87861.1 - 1 23
D87862.1 - 1 23
DQ075334.1 - 1 23
DQ075335.1 - 1 23
DQ075336.1 - 1 23
DQ075337.1 - 1 23
DQ075338.1 - 1 23
DQ075339.1 - 1 23
S82919.1 - 1 23
U43677.1 - 1 28
U43774.1 - 1 28
U56236.1 - 1 33
U56237.1 - 1 33
X92558.1 - 1 23
X92559.1 - 1 23
X92560.1 - 1 23
X92561.1 - 1 23
X92562.1 - 1 23

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
IgA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0