This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CAPNS1 SIRNA Assays ›
Gene Symbol:
CAPNS1
Gene Name:
calpain small subunit 1
Gene Aliases:
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
Chromosome Location:
Chr.19: 36139926 - 36150353 on Build GRCh38
UniGene ID:
Hs.515371
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 212879

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CAPNS1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003962.2 NP_001003962.1 11 1178
NM_001302632.1 NP_001289561.1 11 1085
NM_001302633.1 NP_001289562.1 10 870
NM_001749.3 NP_001740.1 11 1181
XM_005259295.1 XP_005259352.1 10 1157
XM_005259296.1 XP_005259353.1 10 1160
XM_011527359.1 XP_011525661.1 10 811
GenBank mRNA AK289606.1 - 11 1120
AY007141.1 - 11 978
BC000592.2 - 11 999 3347146
BC007779.2 - 11 1009 4301394
BC011903.2 - 10 1175 4120579
BC017308.2 - 11 978 5089022
BC018931.2 - 11 1069 4303926
BC021933.1 - 11 973 4872635
BC023643.2 - 11 971 5017512
BC064998.1 - 11 1086 5785274
BQ891655.1 - 8 642
X04106.1 - 11 1092

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
positive regulation of cell proliferation
regulation of macroautophagy
extracellular matrix disassembly
calcium-dependent cysteine-type endopeptidase activity
calcium ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0