This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PPM1F SIRNA Assays ›
Gene Symbol:
Ppm1f
Gene Name:
protein phosphatase, Mg2+/Mn2+ dependent, 1F
Gene Aliases:
CaMKPase
Chromosome Location:
Chr.11: 88313711 - 88343698 on Build Rnor_6.0
UniGene ID:
Rn.91922
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 201623

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ppm1f
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175755.2 NP_786931.1 6, 7 980
XM_006248674.1 XP_006248736.1 6, 7 1085
XM_008768853.1 XP_008767075.1 6, 7 1031
XM_008768854.1 XP_008767076.1 6, 7 1067
XM_008768855.1 XP_008767077.1 6, 7 1085
GenBank mRNA AB023634.1 - 6, 7 980
BC166854.1 - 6, 7 1085 9098491

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein kinase activity
protein dephosphorylation
positive regulation of gene expression
positive regulation of epithelial cell migration
positive regulation of cell-substrate adhesion
histone dephosphorylation
negative regulation of peptidyl-serine phosphorylation
cellular response to drug
peptidyl-threonine dephosphorylation
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of protein kinase activity by regulation of protein phosphorylation
negative regulation of transcription, DNA-templated
positive regulation of growth
positive regulation of chemotaxis
positive regulation of stress fiber assembly
positive regulation of focal adhesion assembly
intrinsic apoptotic signaling pathway
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
phosphatase activity
calmodulin-dependent protein phosphatase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0