This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MOG SIRNA Assays ›
Gene Symbol:
Mog
Gene Name:
myelin oligodendrocyte glycoprotein
Gene Aliases:
-
Chromosome Location:
Chr.20: 2003575 - 2014284 on Build Rnor_6.0
UniGene ID:
Rn.9687
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 200972

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mog
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022668.2 NP_073159.2 8 1312
XM_006255867.2 XP_006255929.1 7 2312
XM_008772687.1 XP_008770909.1 7 2069
GenBank mRNA BC087717.1 - 8 1312 7317044
M99485.1 - 8 1312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
aging
response to lipid
positive regulation of MyD88-dependent toll-like receptor signaling pathway
response to folic acid
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header