This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PROC SIRNA Assays ›
Gene Symbol:
Proc
Gene Name:
protein C
Gene Aliases:
-
Chromosome Location:
Chr.18: 24918402 - 24928822 on Build Rnor_6.0
UniGene ID:
Rn.91064
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 200474

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Proc
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012803.1 NP_036935.1 6 467
XM_006254526.2 XP_006254588.1 6 539
XM_006254527.2 XP_006254589.1 6 546
XM_008772018.1 XP_008770240.1 6 609
GenBank mRNA BC078879.1 - 5 444 7122006
X64336.1 - 6 467

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

proteolysis
blood coagulation
negative regulation of blood coagulation
negative regulation of apoptotic process
negative regulation of inflammatory response
negative regulation of coagulation
serine-type endopeptidase activity
calcium ion binding
peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0