This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LIPA SIRNA Assays ›
Gene Symbol:
Lipa
Gene Name:
lipase A, lysosomal acid, cholesterol esterase
Gene Aliases:
Chole, Chole2, LAL, Lip1
Chromosome Location:
Chr.1: 252816536 - 252870302 on Build Rnor_6.0
UniGene ID:
Rn.48656
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 200308

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Lipa
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012732.3 NP_036864.3 5, 6 821
XM_006231275.2 XP_006231337.1 5, 6 788
XM_008760321.1 XP_008758543.1 4, 5 684
GenBank mRNA BC072532.1 - 5, 6 821 7099460
S81497.1 - 5, 6 767

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
cytokine production
fatty acid metabolic process
inflammatory response
cell proliferation
lipid catabolic process
sterol metabolic process
lung development
tissue remodeling
homeostasis of number of cells within a tissue
sterol esterase activity
lipase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header