This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC4A1 SIRNA Assays ›
Gene Symbol:
Slc4a1
Gene Name:
solute carrier family 4 (anion exchanger), member 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 90296144 - 90312402 on Build Rnor_6.0
UniGene ID:
Rn.32202
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 200119

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc4a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012651.2 NP_036783.2 9 1112
XM_008767946.1 XP_008766168.1 9 1118
GenBank mRNA AY030082.1 - 6 734
BC085748.1 - 6 1123 7126960

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_011403, BC052419, BC053429, AK172316, X03917, and others.

Gene Ontology Categories:

Function(s) Process(es)

anion transport
chloride transport
bicarbonate transport
regulation of intracellular pH
chloride transmembrane transport
actin binding
inorganic anion exchanger activity
protein C-terminus binding
anion transmembrane transporter activity
bicarbonate transmembrane transporter activity
chloride transmembrane transporter activity
anion:anion antiporter activity
ankyrin binding
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0