This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GCK SIRNA Assays ›
Gene Symbol:
Gck
Gene Name:
glucokinase
Gene Aliases:
GLUKA, RNGK2
Chromosome Location:
Chr.14: 86149146 - 86191576 on Build Rnor_6.0
UniGene ID:
Rn.10447
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199921

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gck
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270850.1 NP_001257779.1 2 197
NM_012565.2 NP_036697.1 2 269
XM_006251179.2 XP_006251241.1 2 1114
GenBank mRNA J04218.1 - 2 228
KJ026953.1 - 2 161
M25807.1 - 2 197
M58759.1 - 2 651
X53588.1 - 2 252

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular glucose homeostasis
glycogen biosynthetic process
fructose 2,6-bisphosphate metabolic process
glucose metabolic process
glycolytic process
NADP metabolic process
positive regulation of cytosolic calcium ion concentration
response to glucose
second-messenger-mediated signaling
positive regulation of insulin secretion
negative regulation of epinephrine secretion
cellular response to insulin stimulus
cellular response to glucose starvation
positive regulation of phosphorylation
glucose homeostasis
regulation of potassium ion transport
cellular response to leptin stimulus
negative regulation of gluconeogenesis
positive regulation of glycogen biosynthetic process
positive regulation of glycolytic process
carbohydrate phosphorylation
regulation of insulin secretion
glucose 6-phosphate metabolic process
detection of glucose
lipid homeostasis
calcium ion import
magnesium ion binding
glucokinase activity
protein binding
ATP binding
glucose binding
fructokinase activity
mannokinase activity
protein phosphatase binding
ADP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0