This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBP1 SIRNA Assays ›
Gene Symbol:
Fbp1
Gene Name:
fructose-1,6-bisphosphatase 1
Gene Aliases:
Fdp
Chromosome Location:
Chr.17: 396175 - 417480 on Build Rnor_6.0
UniGene ID:
Rn.33703
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199906

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fbp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012558.3 NP_036690.2 7 1304
GenBank mRNA BC078894.1 - 7 1286 7114766
BC078895.1 - 7 1304 7134943
J04112.1 - 7 1100
M86240.1 - 7 1190

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
fructose 6-phosphate metabolic process
gluconeogenesis
regulation of gluconeogenesis
dephosphorylation
negative regulation of cell growth
cellular response to insulin stimulus
cellular response to drug
negative regulation of glycolytic process
negative regulation of Ras protein signal transduction
protein homotetramerization
cellular response to magnesium ion
AMP binding
fructose 1,6-bisphosphate 1-phosphatase activity
identical protein binding
metal ion binding
monosaccharide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0