This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATP1A3 SIRNA Assays ›
Gene Symbol:
Atp1a3
Gene Name:
ATPase, Na+/K+ transporting, alpha 3 polypeptide
Gene Aliases:
Atpa1a3
Chromosome Location:
Chr.1: 81852425 - 81881549 on Build Rnor_6.0
UniGene ID:
Rn.87329
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199785

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atp1a3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012506.1 NP_036638.1 12 1624
XM_008758888.1 XP_008757110.1 12 1538
GenBank mRNA FQ212331.1 - 11 1461
M14513.1 - 12 1624
M28648.1 - 11 1478
X05883.1 - 12 1621

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
memory
metabolic process
adult locomotory behavior
visual learning
potassium ion import
ATP hydrolysis coupled proton transport
cellular potassium ion homeostasis
ionotropic glutamate receptor signaling pathway
sodium ion export from cell
response to drug
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
cation-transporting ATPase activity
D1 dopamine receptor binding
metal ion binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0