This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ALDH3A1 SIRNA Assays ›
Gene Symbol:
Aldh3a1
Gene Name:
aldehyde dehydrogenase 3 family, member A1
Gene Aliases:
AHDC, Ahd-c, Aldh, Aldh3
Chromosome Location:
Chr.10: 47490168 - 47499855 on Build Rnor_6.0
UniGene ID:
Rn.105627
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199567

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Aldh3a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031972.1 NP_114178.1 5 844
GenBank mRNA BC070924.1 - 5 877 7096634
J03637.1 - 5 844

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007436, NM_001112725, U12785, BC150677, AF072815, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cellular aldehyde metabolic process
aging
response to nutrient
positive regulation of cell proliferation
response to organic cyclic compound
response to drug
response to glucocorticoid
response to cAMP
oxidation-reduction process
3-chloroallyl aldehyde dehydrogenase activity
aldehyde dehydrogenase (NAD) activity
aldehyde dehydrogenase [NAD(P)+] activity
alcohol dehydrogenase (NADP+) activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0