This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NR1H3 SIRNA Assays ›
Gene Symbol:
Nr1h3
Gene Name:
nuclear receptor subfamily 1, group H, member 3
Gene Aliases:
LXRalpha
Chromosome Location:
Chr.3: 80004130 - 80014197 on Build Rnor_6.0
UniGene ID:
Rn.11209
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199496

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nr1h3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031627.2 NP_113815.2 2, 3 244
XM_006234533.2 XP_006234595.1 3, 4 422
XM_006234534.1 XP_006234596.1 3, 4 363
XM_006234535.2 XP_006234597.1 3, 4 564
GenBank mRNA BC088750.1 - 2, 3 244 7314121
U11685.1 - 2, 3 251

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
lipid metabolic process
insulin receptor signaling pathway
positive regulation of triglyceride biosynthetic process
positive regulation of receptor biosynthetic process
positive regulation of cholesterol efflux
negative regulation of cholesterol storage
intracellular receptor signaling pathway
positive regulation of cellular protein metabolic process
negative regulation of lipid transport
positive regulation of cholesterol transport
response to progesterone
positive regulation of toll-like receptor 4 signaling pathway
cholesterol homeostasis
negative regulation of macrophage activation
apoptotic cell clearance
steroid hormone mediated signaling pathway
cellular lipid metabolic process
positive regulation of fatty acid biosynthetic process
negative regulation of proteolysis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of pinocytosis
negative regulation of inflammatory response
positive regulation of lipoprotein lipase activity
lipid homeostasis
sterol homeostasis
triglyceride homeostasis
cellular response to lipopolysaccharide
negative regulation of pancreatic juice secretion
negative regulation of secretion of lysosomal enzymes
regulation of cholesterol homeostasis
positive regulation of cholesterol homeostasis
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
zinc ion binding
sterol response element binding
sequence-specific DNA binding
transcription regulatory region DNA binding
retinoid X receptor binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0