This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YWHAE SIRNA Assays ›
Gene Symbol:
Ywhae
Gene Name:
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilon
Gene Aliases:
14-3-3e
Chromosome Location:
Chr.10: 63884338 - 63921709 on Build Rnor_6.0
UniGene ID:
Rn.4225
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199446

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ywhae
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031603.1 NP_113791.1 6 1604
GenBank mRNA FQ215270.1 - 4 1262
FQ220393.1 - 6 1610
M84416.1 - 6 1584
U53882.1 - 6 1604

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_006761, U43430, BC000179, U43399, U54778, and others.

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
protein targeting
signal transduction
substantia nigra development
hippocampus development
cerebral cortex development
negative regulation of protein dephosphorylation
regulation of membrane repolarization
negative regulation of protein serine/threonine kinase activity
regulation of potassium ion transmembrane transporter activity
negative regulation of peptidyl-serine dephosphorylation
protein binding
protein kinase C inhibitor activity
potassium channel regulator activity
enzyme binding
protein domain specific binding
MHC class II protein complex binding
protein complex binding
histone deacetylase binding
ion channel binding
poly(A) RNA binding
protein heterodimerization activity
phosphoserine binding
phosphoprotein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header