This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC37A4 SIRNA Assays ›
Gene Symbol:
Slc37a4
Gene Name:
solute carrier family 37 (glucose-6-phosphate transporter), member 4
Gene Aliases:
G6pt1
Chromosome Location:
Chr.8: 48716939 - 48723024 on Build Rnor_6.0
UniGene ID:
Rn.1592
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199418

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Slc37a4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031589.2 NP_113777.2 2 354
XM_008766151.1 XP_008764373.1 1 84
XM_008766152.1 XP_008764374.1 1 180
XM_008766153.1 XP_008764375.1 1 84
GenBank mRNA AF080468.1 - 1 184
BC070889.1 - 2 354 7098680

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

neutrophil homeostasis
cytokine production
myeloid progenitor cell differentiation
glycogen metabolic process
lactate metabolic process
triglyceride metabolic process
cellular calcium ion homeostasis
response to nutrient
steroid metabolic process
response to glucose
response to organic cyclic compound
glucose-6-phosphate transport
neutrophil chemotaxis
negative regulation of chemokine production
post-embryonic hemopoiesis
glucose homeostasis
cholesterol homeostasis
positive regulation of catalytic activity
respiratory burst
transmembrane transport
glucose-6-phosphate transmembrane transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0