This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AZIN1 SIRNA Assays ›
Gene Symbol:
Azin1
Gene Name:
antizyme inhibitor 1
Gene Aliases:
Oazi, Oazin
Chromosome Location:
Chr.7: 77345671 - 77372393 on Build Rnor_6.0
UniGene ID:
Rn.6290
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199242

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Azin1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301702.1 NP_001288631.1 4 803
NM_022585.2 NP_072107.2 5 961
XM_006241568.2 XP_006241630.1 5 934
XM_006241569.2 XP_006241631.1 4 798
XM_008765457.1 XP_008763679.1 4 798
GenBank mRNA BC061550.1 - 2 199 6918922
D50734.1 - 4 596
D89983.1 - 5 909

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

polyamine biosynthetic process
negative regulation of protein catabolic process
positive regulation of catalytic activity
negative regulation of catalytic activity
positive regulation of polyamine transmembrane transport
catalytic activity
ornithine decarboxylase inhibitor activity
ornithine decarboxylase activator activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0