This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DLG4 SIRNA Assays ›
Gene Symbol:
Dlg4
Gene Name:
discs, large homolog 4 (Drosophila)
Gene Aliases:
Dlgh4, PSD95, Sap90
Chromosome Location:
Chr.10: 56625630 - 56655543 on Build Rnor_6.0
UniGene ID:
Rn.9765
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 199086

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dlg4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019621.1 NP_062567.1 9 892
XM_006246680.2 XP_006246742.1 11 1268
XM_006246681.2 XP_006246743.1 11 1263
XM_006246682.2 XP_006246744.1 11 1117
XM_006246683.2 XP_006246745.1 11 1025
XM_006246684.2 XP_006246746.1 11 1016
XM_006246685.2 XP_006246747.1 9 978
XM_006246686.2 XP_006246748.1 9 1038
XM_006246687.2 XP_006246749.1 9 938
XM_006246688.2 XP_006246750.1 8 3139
XM_006246689.2 XP_006246751.1 8 881
XM_006246691.2 XP_006246753.1 8 872
XM_008767809.1 XP_008766031.1 11 1369
XM_008767810.1 XP_008766032.1 8 756
GenBank mRNA M96853.1 - 9 892
X66474.1 - 9 1037

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of receptor internalization
protein complex assembly
positive regulation of cytosolic calcium ion concentration
synaptic vesicle maturation
social behavior
protein localization to synapse
locomotory exploration behavior
neuronal ion channel clustering
establishment of protein localization
establishment or maintenance of epithelial cell apical/basal polarity
nucleotide phosphorylation
regulation of long-term neuronal synaptic plasticity
positive regulation of synaptic transmission
neuromuscular process controlling balance
dendritic spine morphogenesis
vocalization behavior
AMPA glutamate receptor clustering
receptor localization to synapse
regulation of N-methyl-D-aspartate selective glutamate receptor activity
positive regulation of excitatory postsynaptic potential
regulation of grooming behavior
guanylate kinase activity
receptor binding
protein binding
protein C-terminus binding
protein kinase binding
protein phosphatase binding
PDZ domain binding
beta-1 adrenergic receptor binding
D1 dopamine receptor binding
P2Y1 nucleotide receptor binding
protein complex binding
acetylcholine receptor binding
glutamate receptor binding
ionotropic glutamate receptor binding
scaffold protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0