This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RALGDS SIRNA Assays ›
Gene Symbol:
Ralgds LOC102554500
Gene Name:
ral guanine nucleotide dissociation stimulator
ral guanine nucleotide dissociation stimulator-like
Gene Aliases:
Rgds
-
Chromosome Location:
Chr.3: 7090134 - 7130548 on Build Rnor_6.0
Chromosome Location:
Chr.3: 4987820 - 4989204 on Build Rnor_6.0
UniGene ID:
Rn.40174
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 198831

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ralgds
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019250.2 NP_062123.2 6 1289
XM_006233750.2 XP_006233812.1 6 1834
XM_006233751.2 XP_006233813.1 6 3767
XM_006233752.2 XP_006233814.1 6 1347
XM_006233753.2 XP_006233815.1 6 1311
XM_008761632.1 XP_008759854.1 6 1612
GenBank mRNA L07925.1 - 6 1289
Gene Symbol: LOC102554500
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006233730.2 XP_006233792.1 2 880

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

small GTPase mediated signal transduction
positive regulation of GTPase activity
regulation of catalytic activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Ral guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0