This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GALR2 SIRNA Assays ›
Gene Symbol:
Galr2 LOC103690156
Gene Name:
galanin receptor 2
galanin receptor type 2
Gene Aliases:
-
-
Chromosome Location:
Chr.10: 105156802 - 105158967 on Build Rnor_6.0
Chromosome Location:
Chr.10: 105234247 - 105241704 on Build Rnor_6.0
UniGene ID:
Rn.9789
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 198656

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Galr2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019172.4 NP_062045.1 2 722
GenBank mRNA AF008548.1 - 2 843
AF010318.1 - 2 722
U94322.1 - 2 1192
Y15248.1 - 2 702
Gene Symbol: LOC103690156
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768432.1 XP_008766654.1 1744

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
negative regulation of adenylate cyclase activity
positive regulation of cytosolic calcium ion concentration
neuropeptide signaling pathway
neuron projection development
inositol phosphate metabolic process
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol metabolic process
positive regulation of large conductance calcium-activated potassium channel activity
galanin receptor activity
peptide hormone binding
neuropeptide binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0