This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GRIA4 SIRNA Assays ›
Gene Symbol:
Gria4
Gene Name:
glutamate receptor, ionotropic, AMPA 4
Gene Aliases:
GluA4, GluR-D, GluR4
Chromosome Location:
Chr.8: 1548145 - 2045817 on Build Rnor_6.0
UniGene ID:
Rn.10938
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198414

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gria4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113184.1 NP_001106655.1 3 636
NM_001113185.1 NP_001106656.1 3 636
NM_017263.2 NP_058959.2 3 636
GenBank mRNA BC093608.1 - 3 608
M36421.1 - 2 186
M38063.1 - 2 186
M85037.1 - 3 347

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chemical synaptic transmission
ion transmembrane transport
ionotropic glutamate receptor signaling pathway
regulation of synapse structure or activity
modulation of synaptic transmission
positive regulation of synaptic transmission, glutamatergic
response to fungicide
ionotropic glutamate receptor activity
AMPA glutamate receptor activity
extracellular-glutamate-gated ion channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0