This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYH7 SIRNA Assays ›
Gene Symbol:
Myh7
Gene Name:
myosin, heavy chain 7, cardiac muscle, beta
Gene Aliases:
Myhcb
Chromosome Location:
Chr.15: 33634420 - 33657761 on Build Rnor_6.0
UniGene ID:
Rn.225886
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198362

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Myh7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017240.2 NP_058936.1 1 112
XM_006251951.2 XP_006252013.1 3 262
GenBank mRNA X15939.1 - 1 114

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080728, AK147031, BC158018, AK157742, DQ231556, and others.

Gene Ontology Categories:

Function(s) Process(es)

response to reactive oxygen species
regulation of the force of heart contraction
regulation of heart rate
muscle contraction
striated muscle contraction
adult heart development
metabolic process
transition between fast and slow fiber
muscle filament sliding
ventricular cardiac muscle tissue morphogenesis
microfilament motor activity
actin binding
calmodulin binding
ATP binding
ATPase activity
actin-dependent ATPase activity
protein homodimerization activity
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0