This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHRNE SIRNA Assays ›
Gene Symbol:
Chrne
Gene Name:
cholinergic receptor, nicotinic, epsilon (muscle)
Gene Aliases:
-
Chromosome Location:
Chr.10: 57238960 - 57247632 on Build Rnor_6.0
UniGene ID:
Rn.10301
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198257

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chrne
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017194.1 NP_058890.1 6, 7 641
XM_006246678.2 XP_006246740.1 7, 8 759
XM_008767807.1 XP_008766029.1 7, 8 742
XM_008767808.1 XP_008766030.1 8, 9 1408
GenBank mRNA X13252.1 - 6, 7 641
X74836.1 - 6, 7 641

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_009603, X55718.

Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle contraction
cation transport
regulation of membrane potential
cation transmembrane transport
acetylcholine-activated cation-selective channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0