This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MEOX2 SIRNA Assays ›
Gene Symbol:
Meox2
Gene Name:
mesenchyme homeobox 2
Gene Aliases:
-
Chromosome Location:
Chr.6: 56625768 - 56689195 on Build Rnor_6.0
UniGene ID:
Rn.34890
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198148

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Meox2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017149.1 NP_058845.1 3 1611
GenBank mRNA Z17223.1 - 3 1611

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
somite specification
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
skeletal muscle tissue development
positive regulation of transcription from RNA polymerase II promoter
palate development
limb development
neuron death
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0