This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FDX1 SIRNA Assays ›
Gene Symbol:
Fdx1
Gene Name:
ferredoxin 1
Gene Aliases:
-
Chromosome Location:
Chr.8: 56373729 - 56393199 on Build Rnor_6.0
UniGene ID:
Rn.6946
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198094

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fdx1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017126.1 NP_058822.1 4 554
GenBank mRNA D50436.1 - 4 554
DQ255900.1 - 2 158

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007996, BC099518, BC132173, AK004879, BC132175, and others.

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
cholesterol metabolic process
adrenal gland development
response to gonadotropin
hormone biosynthetic process
cellular response to leptin stimulus
response to leptin
regulation of catalytic activity
positive regulation of oxidoreductase activity
NADPH oxidation
cellular response to organic cyclic compound
cellular response to epinephrine stimulus
iron ion binding
cholesterol monooxygenase (side-chain-cleaving) activity
electron carrier activity
NADPH-adrenodoxin reductase activity
enzyme binding
metal ion binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0