This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CSF3 SIRNA Assays ›
Gene Symbol:
Csf3
Gene Name:
colony stimulating factor 3 (granulocyte)
Gene Aliases:
Gcsf
Chromosome Location:
Chr.10: 86616708 - 86619160 on Build Rnor_6.0
UniGene ID:
Rn.53973
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 198041

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Csf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017104.1 NP_058800.1 5 831
XM_006247265.2 XP_006247327.2 5 904
GenBank mRNA U37101.1 - 5 831

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune response
positive regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
positive regulation of actin filament polymerization
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription factor import into nucleus
positive regulation of myeloid cell differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of peptidyl-tyrosine phosphorylation
positive regulation of protein kinase B signaling
cellular response to lipopolysaccharide
cellular response to cytokine stimulus
negative regulation of neuron death
positive regulation of actin cytoskeleton reorganization
cytokine activity
growth factor activity
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0