This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PDE4A SIRNA Assays ›
Gene Symbol:
Pde4a
Gene Name:
phosphodiesterase 4A, cAMP-specific
Gene Aliases:
PHOSA
Chromosome Location:
Chr.8: 22189533 - 22234036 on Build Rnor_6.0
UniGene ID:
Rn.91357
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197751

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pde4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013101.3 NP_037233.3 9 1230
GenBank mRNA AF110461.2 - 9 922
L27057.1 - 9 1230
L27062.1 - 3 760
L36467.1 - 1120
M25348.1 - 2 190
M26715.1 - 3 494
M26716.1 - 3 398
M28411.1 - 2 190

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_019798, NM_183408, BC139405, AF208021, BC139408, and others.

Gene Ontology Categories:

Function(s) Process(es)

cAMP catabolic process
signal transduction
sensory perception of smell
metabolic process
regulation of protein kinase A signaling
cellular response to drug
response to drug
regulation of cAMP-mediated signaling
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
protein binding
cAMP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0