This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD81 SIRNA Assays ›
Gene Symbol:
Cd81
Gene Name:
Cd81 molecule
Gene Aliases:
Tapa1
Chromosome Location:
Chr.1: 216237697 - 216253311 on Build Rnor_6.0
UniGene ID:
Rn.1975
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197712

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cd81
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013087.1 NP_037219.1 2 387
GenBank mRNA BC060583.1 - 2 218 6918296
FQ231991.1 - 2 373
FQ232886.1 - 2 375
U19894.1 - 2 387

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
phosphatidylinositol biosynthetic process
protein localization
positive regulation of cell proliferation
response to wounding
positive regulation of B cell proliferation
regulation of growth
regulation of cell proliferation
positive regulation of 1-phosphatidylinositol 4-kinase activity
phosphatidylinositol metabolic process
positive regulation of peptidyl-tyrosine phosphorylation
regulation of cell motility
protein binding
MHC class II protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0