This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other YWHAZ SIRNA Assays ›
Gene Symbol:
Ywhaz
Gene Name:
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zeta
Gene Aliases:
14-3-3z
Chromosome Location:
Chr.7: 75573546 - 75598295 on Build Rnor_6.0
UniGene ID:
Rn.1292
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197525

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ywhaz
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013011.3 NP_037143.2 2 183
XM_006241529.2 XP_006241591.1 3 736
GenBank mRNA BC070941.1 - 2 183 7099670
BC094305.1 - 2 488 7313193
D17615.1 - 2 250
D30740.1 - 2 163
U37252.1 - 2 184

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

histamine secretion by mast cell
protein targeting
protein targeting to mitochondrion
regulation of cell death
response to drug
establishment of Golgi localization
Golgi reassembly
protein binding
transcription factor binding
protein kinase binding
protein domain specific binding
protein complex binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0