This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BCAN SIRNA Assays ›
Gene Symbol:
Bcan
Gene Name:
brevican
Gene Aliases:
ALPBRE, BEHAB
Chromosome Location:
Chr.2: 187359674 - 187372652 on Build Rnor_6.0
UniGene ID:
Rn.10315
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197291

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Bcan
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033665.1 NP_001028837.1 5 917
NM_012916.2 NP_037048.2 5 917
XM_006232595.2 XP_006232657.1 5 1998
XM_006232596.2 XP_006232658.1 5 901
XM_006232597.2 XP_006232659.1 5 719
GenBank mRNA BC094307.1 - 1 1697
U37142.1 - 5 721
X79881.1 - 5 771
X86406.1 - 5 771
Z28366.1 - 5 912

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
cell adhesion
central nervous system development
hippocampus development
regulation of neuronal synaptic plasticity
extracellular matrix structural constituent
hyaluronic acid binding
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0