This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANXA1 SIRNA Assays ›
Gene Symbol:
Anxa1
Gene Name:
annexin A1
Gene Aliases:
Anx1, p35
Chromosome Location:
Chr.1: 237893983 - 237921165 on Build Rnor_6.0
UniGene ID:
Rn.1792
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197266

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Anxa1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012904.2 NP_037036.1 13 1060
XM_006231159.1 XP_006231221.1 14 1228
XM_008760295.1 XP_008758517.1 13 1147
GenBank mRNA BC061710.1 - 13 1058 5598112
FQ213489.1 - 13 1075
FQ215033.1 - 13 1075
FQ216134.1 - 13 1075
FQ228716.1 - 13 1075
FQ229071.1 - 13 1075
FQ229198.1 - 12 996
M19967.1 - 12 1027
Y00446.1 - 13 1061

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010730, AK150564, AK150718, EU684130, BC002289, and others.

Gene Ontology Categories:

Function(s) Process(es)

DNA strand renaturation
neutrophil homeostasis
signal transduction
cell surface receptor signaling pathway
response to hormone
response to X-ray
response to organic cyclic compound
peptide cross-linking
insulin secretion
keratinocyte differentiation
prostate gland development
endocrine pancreas development
positive regulation of vesicle fusion
positive regulation of prostaglandin biosynthetic process
response to corticosteroid
response to estradiol
DNA duplex unwinding
positive regulation of neutrophil apoptotic process
DNA rewinding
gliogenesis
regulation of cell proliferation
response to drug
positive regulation of apoptotic process
response to peptide hormone
estrous cycle
alpha-beta T cell differentiation
arachidonic acid secretion
negative regulation of protein secretion
response to glucocorticoid
cellular response to hydrogen peroxide
hepatocyte differentiation
prolactin secretion
response to interleukin-1
cellular response to glucocorticoid stimulus
neutrophil clearance
positive regulation of G1/S transition of mitotic cell cycle
negative regulation of phospholipase A2 activity
negative regulation of interleukin-8 secretion
single-stranded DNA binding
single-stranded RNA binding
helicase activity
structural molecule activity
calcium ion binding
phospholipid binding
calcium-dependent phospholipid binding
phospholipase A2 inhibitor activity
protein binding, bridging
double-stranded DNA-dependent ATPase activity
annealing helicase activity
protein homodimerization activity
calcium-dependent protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0