This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NFYA SIRNA Assays ›
Gene Symbol:
Nfya
Gene Name:
nuclear transcription factor-Y alpha
Gene Aliases:
CBF-A
Chromosome Location:
Chr.9: 14551752 - 14577002 on Build Rnor_6.0
UniGene ID:
Rn.10747
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197162

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nfya
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012865.1 NP_036997.1 4 352
XM_006244409.2 XP_006244471.1 4 359
XM_006244410.1 XP_006244472.1 4 355
XM_006244411.2 XP_006244473.1 4 349
XM_006244412.2 XP_006244474.1 4 346
XM_006244413.1 XP_006244475.1 3 267
XM_008766839.1 XP_008765061.1 3 255
GenBank mRNA FQ210330.1 - 3 272
FQ223023.1 - 4 364
M34238.1 - 4 352

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
rhythmic process
regulation of stem cell population maintenance
positive regulation of stem cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription cofactor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
transcription regulatory region DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0