This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CACNB3 SIRNA Assays ›
Gene Symbol:
Cacnb3
Gene Name:
calcium channel, voltage-dependent, beta 3 subunit
Gene Aliases:
CACH3B
Chromosome Location:
Chr.7: 140311140 - 140324902 on Build Rnor_6.0
UniGene ID:
Rn.2808
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 197067

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cacnb3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012828.2 NP_036960.1 3 294
XM_006257329.2 XP_006257391.1 3 366
XM_006257330.2 XP_006257392.1 3 775
GenBank mRNA CB739910.1 - 3 420
M88751.1 - 3 298

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001206917, NM_001206916, NM_000725, AK304537, BI667991, and others.
This siRNA also targets the following transcripts in Mouse: NM_001286226, NM_001044741, NM_007581, U20372, AK197162, and others.

Gene Ontology Categories:

Function(s) Process(es)

calcium ion transport
chemical synaptic transmission
neuromuscular junction development
T cell receptor signaling pathway
calcium ion transmembrane transport
regulation of voltage-gated calcium channel activity
voltage-gated calcium channel activity
protein binding
high voltage-gated calcium channel activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0