This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DLD SIRNA Assays ›
Gene Symbol:
Dld
Gene Name:
dihydrolipoamide dehydrogenase
Gene Aliases:
-
Chromosome Location:
Chr.6: 50597677 - 50618694 on Build Rnor_6.0
UniGene ID:
Rn.86962
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID 196972

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dld
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199385.2 NP_955417.1 9 878
XM_008764590.1 XP_008762812.1 9 895
GenBank mRNA BC062069.1 - 9 882 5599215
FM046215.1 - 1 48
FM093110.1 - 4 456
FQ215299.1 - 9 885

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

acetyl-CoA biosynthetic process from pyruvate
2-oxoglutarate metabolic process
mitochondrial electron transport, NADH to ubiquinone
proteolysis
gastrulation
aging
lipoate metabolic process
regulation of membrane potential
cell redox homeostasis
sperm capacitation
detoxification of mercury ion
dihydrolipoamide metabolic process
oxidation-reduction process
dihydrolipoyl dehydrogenase activity
mercury (II) reductase activity
lipoamide binding
mercury ion binding
flavin adenine dinucleotide binding
NADP binding
NAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0